Hydrolysis unit, HU 6
Leverandør: VELP SCIENTIFIC
F30300150
VELPF30300150EA 47525 NOK
VELPF30300150
Hydrolysis unit, HU 6
Ekstraktorer Systemer til fiberbestemmelse
Convenient, reliable and safe acid and basic hydrolysis unit, for sample preparation prior to extraction for total fat determination.
  • Fast and precise heating with 0,5 °C temperature stability and homogeneity
  • Convenient operation saving user time
  • Faster and enhanced filtration thanks to valves that allow selective interruption of the vacuum at each position
  • Continuous filtration and rinsing six samples in parallel
  • Intuitive digital interface
  • Low energy consumption
The HU 6 hydrolysis unit is the ideal solution for treatment prior to solvent extractionMore Product Information