Portable turbidity meter
Leverandør: VELP SCIENTIFIC
R109B12150
VELPR109B12150EA 14825 NOK
VELPR109B12150
Portable turbidity meter
Turbiditetsmålere
The TB1 can measure the turbidity in aqueous samples for industrial and environmental fields.
  • Intuitive and easy calibration
  • Results provided in a few seconds
  • Subsequent analysis is more precise
Sertifiseringer: NIST traceable calibration. ISO 7027 compliant nephelometric method (90°).
Leveringsinformasjon: Supplied with calibration standards, three vials, cloth, silicone oil and batteries.