Ceramic Magnetic Hotplate Stirrers, AREC 4 Digital
Leverandør: VELP SCIENTIFIC
SC20500671 SA20500671 SB20500671 F20500671
VELPSC20500671EA 7925 NOK
VELPSC20500671 VELPSA20500671 VELPSB20500671 VELPF20500671
Ceramic Magnetic Hotplate Stirrers, AREC 4 Digital
Omrørere Magnetrørere med varmeplate
Powerful digital hotplate stirrer that features a 10 cm (4") ceramic plate and guarantees precise thermoregulation of the media, high safety and maximum reliability. The AREC 4 Digital has an aluminium body and technopolymer housing, it offers precise temperature control, advanced stirring and heating performance, improved safety features, and ease of use. The compact footprint makes it an indispensable solution for laboratories with limited bench space.
  • Advanced heating plate technology ensures temperature homogeneity and stability up to 550 °C
  • Precise