Ultra Chemical Resistant Tubing for Tygon® Ink 1000
Leverandør: Saint Gobain Life Sciences
A0C1S1520
VERNA0C1S1520EA 0 NOK
VERNA0C1S1520
Ultra Chemical Resistant Tubing for Tygon® Ink 1000
Slanger Overføringsslange
For UV inks in particular, UV blocking properties is an industry standard. Tygon® Ink 1000 has shown over an accelerated testing period that it can maintain the ink integrity with our patented technology, making it an ideal candidate for the UV market.
  • Outstanding chemical resistance to small acrylic monomers
  • Flexibility
  • UV blocking
  • Low extractable levels
  • Long service life
Chemical Resistance: In the ink transfer space, chemical compatibility and flexibility are two of the most important criteria for ensuring satisfactory performance. Tygon® Ink 1000More Product Information