Tube clamps, Pure-Fit® TC
Leverandør: Saint Gobain Life Sciences
PFTC375 PFTC750 PFTC750PVDF PFTC375PVDF
VERNPFTC375EA 2600 NOK
VERNPFTC375 VERNPFTC750 VERNPFTC750PVDF VERNPFTC375PVDF
Tube clamps, Pure-Fit® TC
Klemmer Slangeklemmer
Pure-Fit® TC tube clamps represent a new generation of clamping technology for the pharmaceutical industry.
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Custom colours and identification available
  • Autoclavable and sterilisable (autoclaving not recommended for PP clamps)
They feature an outer body with no sharp edges, eliminating the potential for snags, scratches and punctures; a unique press-down lockingMore Product Information