Sani-Tech® STHT–C Tubings, Silicone, Platinum Cured
Leverandør: Saint Gobain Life Sciences
STHT-C-187-2F STHT-C-187-2H STHT-C-250-3F STHT-C-250-3H STHT-C-312-4F STHT-C-375-2H STHT-C-375-4F STHT-C-375-4H HSTHT-C-1000-6
VERNSTHT-C-187-2FEA 67550 NOK
VERNSTHT-C-187-2F VERNSTHT-C-187-2H VERNSTHT-C-250-3F VERNSTHT-C-250-3H VERNSTHT-C-312-4F VERNSTHT-C-375-2H VERNSTHT-C-375-4F VERNSTHT-C-375-4H VERNHSTHT-C-1000-6
Sani-Tech® STHT–C Tubings, Silicone, Platinum Cured
Slanger Overføringsslange
STHT®-C silicone tubing is an ultra-pure, biopharmaceutical-grade tubing which resists temperature extremes, ozone, corona, radiation, moisture, compression sets, weathering and chemical attack and imparts no taste or odours to fluids transported within it.
  • Unsurpassed surface smoothness
  • Ultra-flexible
  • Temperature resistance from –62 to 260 °C
Permanently laser marked with part number and lot code for total product traceability. Used in sterile fill and transfers, drug delivery, media processing, sanitary peristaltic pump tubing, laboratory use, cellMore Product Information