Cleaning cloths, MicroTuff Plus
Leverandør: FHP VILEDA SCS
111955
VILE111955EA 0 NOK
VILE111955
Cleaning cloths, MicroTuff Plus
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
MicroTuff plus is a high quality warp knitted textile microfiber cloth made of 80% polyester and 20% polyamide. It removes greasy finger marks and stubborn dirt without the use of chemicals. MicroTuff plus can be used in areas with high dirt level, where high volume and dirt pick up are the primary requirements.
  • Color-coded: Cleaning process according to hygiene standards and HACCP principles can be applied
  • Even after several washing cycles the appearance of MicroTuff plus is in line with the high demands of professional cleaning
  • Can be used as a