Cleaning cloths, GP Extra
Leverandør: FHP VILEDA SCS
115090
VILE115090EA 300 NOK
VILE115090
Cleaning cloths, GP Extra
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
Economical cloths with non woven structure for general purpose wet and damp cleaning tasks.
  • Non woven structure: Growth of bacteria and germs is greatly reduced
  • Easily lifts dirt from surfaces
  • High fibre density greatly improves absorbency
  • Easy to rinse
  • Washable at 95 °C