Desiccators
Leverandør: VITLAB
326294
VITL326294EA 9950 NOK
VITL326294
Desiccators
Eksikatorer Eksikator, plast
Ideal dessicators for use in educational laboratories.
  • Lid made from PC is sealed with a neoprene gasket
  • Lower portion made from PP can be filled with desiccants
  • Materials to be dried can be placed on a perforated disc made of PP