Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Leverandør: Merck
VT000036-10EA
VT000036-10EAEA 1550 NOK
VT000036-10EA
Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Standarder Biologiskestandarder
Derived from CECT® strains.
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.