Legionella pneumophila (Serogroup 1) WDCM 00205 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Leverandør: Merck
VT002057-10EA
VT002057-10EAEA 1600 NOK
VT002057-10EA
Legionella pneumophila (Serogroup 1) WDCM 00205 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Standarder Biologiskestandarder
Derived from CECT® strains.
  • Origin Strain No. CECT® 8734
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.