VWR® Rota® pH Indicator Papers
Leverandør: VWR Chemicals
VWRC109526.0005EA 890 NOK
VWRC109526.0005
VWR® Rota® pH Indicator Papers
Testpapir pH-indikatorstrips
These rolls (4,8 m × 10 mm) have only 1 colour zone and are useful where high precision of measurement is not necessary.