VWR®, Laboratory Bottles, Narrow Neck
Leverandør: VWR Collection
VWRI215-1592-EEA 910 NOK
VWRI215-1592-E VWRI215-1593-E VWRI215-1594-E VWRI215-1595-E VWRI215-1596-E
VWR®, Laboratory Bottles, Narrow Neck
Flasker Glassflasker
Borosilicate 3.3 glass, without screw cap and pouring ring.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Economical alternative for a wide range of laboratory applications. Round, with GL 45 thread and white graduations.
Sertifiseringer: ISO 4796
Bestillingsinformasjon: PP screw cap and pouring ring must be ordered separately.