VWR® Bottles, Water Sampling
Leverandør: VWR Collection
331-0286EA 8575 NOK
331-0286 VWRI331-0287.1 VWRI331-0288.1 VWRI331-0283.1 VWRI331-0278.1 VWRI331-0279.1 VWRI331-0280.1 VWRI331-0281.1 VWRI331-0282.1 VWRI331-0286.1 VWRI331-0287_U VWRI331-0288_U
VWR® Bottles, Water Sampling
Flasker Vannprøveflasker
PET, virgin, food grade with PP caps.
  • Irradiated - sterile
  • Different coloured caps for easy indentification of test
  • Tamper-evident closure to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Tamper-evident screw caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Square bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.