VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Leverandør: VWR Collection
VWRI331-0301.1EA 1350 NOK
VWRI331-0301.1
VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Flasker Plastflasker
PET, clear, with blue screw cap.
  • Graduated
  • Tamper evident cap (fitted)
  • Lightweight and robust
  • Square
Perfect for sample collection, storage and transport.