Accessories for VWR® 211DS, Personal Shaking Incubators
Leverandør: VWR Collection
VWRIS2031-13EA 5800 NOK
VWRIS2031-13
Accessories for VWR® 211DS, Personal Shaking Incubators
Inkubatorer Inkubatorer, standard
A full range of accessories is available to complement the 211DS. The 211DS includes three shelves (one is a half-size shelf, allowing tall vessels to be accommodated). Two shaking platforms (sold separately) are available for the 211DS; the flask platform is predrilled to accept a variety of different sized clamps while the flat platform has a nonslip rubber mat for holding dishes and plates in place.