VWR® Bottles, Wide Neck, Square, with Screw Caps
Leverandør: LAB3 OWN PRODUCTION
VWRLSTL102EA 1100 NOK
VWRLSTL102 VWRLSTL100 VWRLSTL101.1
VWR® Bottles, Wide Neck, Square, with Screw Caps
Flasker Plastflasker
PET, clear, with screw cap.
  • Good chemical resistance
  • With or without label available
Ideal for packaging, storing and transporting liquids, pastes and powders. Wide mouth for easy filling, emptying and cleaning.