Trietylamin ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®

Leverandør: Avantor
Danger


Management of Change (MOC) category = N

W635-07EA 1775 NOK
W635-07
Trietylamin ≥99% (by GC), BAKER ANALYZED®, J.T.Baker®
Trietylamin

Lot analysis on label.

Formel: (C₂H₅)₃N
Kokepunkt: 90 °C (1013 hPa)
Smeltepunkt: –115 °C
Tetthet: 0,729 g/cm³ (20 °C)
Flash punkt: –11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009051
CAS-nummer: 121-44-8
EINECS: 204-469-4
FN: 1296
ADR: 3,II
Merck Indeks: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 99 %
Assay ((C₂H₅)₃N) (by GC, corrected for water) ≥ 99 %
Color (APHA) ≤ 15
Water (H₂O) ≤ 1 %
Identification (by IR) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR