BioCleanPlate Wako™ Microplates
Leverandør: FUJIFILM WAKO CHEMICALS
293-35221
WAKF293-35221EA 5875 NOK
WAKF293-35221
BioCleanPlate Wako™ Microplates
Mikroplater Deepwell mikroplater
The BioCleanPlate Wako™ are endotoxin-free microplates that are individually wrapped to reduce the chance of contamination and are perfect for LAL testing.
  • Endotoxin-free 96-well microplate (<0,01 EU/plate)
  • Individually packaged, ideal for use in clean rooms
  • Packaging film features dust-free protection
  • Plates are designed for use with microplate readers
When used in conjunction with BioCleanTip Wako® Extend S allows for aliquoting multi-test reagent into Microplates accurately and precisely avoiding error or bubbles.