Laboratory blenders
Leverandør: WARING BLENDOR
CB15E CB15TE
WARICB15EEA 0 NOK
WARICB15E WARICB15TE
Laboratory blenders
Homogenisatorer Blendere, miksere
The CB15 series blenders are designed specifically for processing large quantities of free flowing material with a high liquid content. This range of 3-speed blenders has an integral pulse button and is fitted with a powerful 3,75 hp motor encased within a die-cast epoxy coated motor housing.
  • Easy to move and fitted with full traction rubber feet for greater stability
  • Easy-to-clean electronic keypad
  • Slow Start™ blending mechanism
  • Stainless steel container jar with two handles
  • Smaller 2, 1 litre and mini-containers can be used with the aid of the AD