Standard cutter
Leverandør: WEDO WERNER DORSCH
785018
WEDO785018EA 270 NOK
WEDO785018
Standard cutter
Kniver
Heavy duty cutter in ABS housing with break-off blade.
  • With ribbed rubber surface to improve grip for safety
  • Rigid all metal carrier for more safety
  • Screw clamp secures the blade
  • Blade break off device is included
Leveringsinformasjon: Supplied with three blades on blister card and break off device.