Qualitative filter papers, standard grades, grade 597L, Whatman™
Leverandør: Whatman products (Cytiva)
10312070
WHAT10312070EA 3900 NOK
WHAT10312070
Qualitative filter papers, standard grades, grade 597L, Whatman™
Filter Filterpapir Kvalitativt filterpapir
These are cellulose qualitative filter papers with low fat content.