Application specific filter papers, grade 1450CV, Whatman™
Leverandør: Whatman products (Cytiva)
10313209
WHAT10313209EA 140 NOK
WHAT10313209
Application specific filter papers, grade 1450CV, Whatman™
Filter Filterpapir Kvalitativt filterpapir
Cellulose filter paper for identification of undissolved dyes in the textile industry.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Vekt 120 g/m²
Filtreringshastighet 30 s (Herzberg)