Application specific filter papers, grade 3459, Whatman™
Leverandør: Whatman products (Cytiva)
10316619
WHAT10316619EA 5700 NOK
WHAT10316619
Application specific filter papers, grade 3459, Whatman™
Filter Filterpapir Kvalitativt filterpapir
Creped cellulose filter papers with good retentivity at a relatively high filtration speed. They are used for the clarifying filtration of dried beet pulp extracts or beet juice after the addition of lead acetate for subsequent polarimetric sugar determination. They are specifically designed for the Venema unit (lead acetate method).
Bestill nå
SPESIFIKASJONER
Nr. 3459
Environmentally Preferable
Vekt 75 g/m²
Tykkelse 300 µm
Filtreringshastighet 55 s (Herzberg)