Application specific filter papers, grade 0048, Whatman™
Leverandør: Whatman products (Cytiva)
10348903
WHAT10348903EA 5175 NOK
WHAT10348903
Application specific filter papers, grade 0048, Whatman™
Filter Filterpapir Kvalitativt filterpapir
Filter mat made from a mixture of cellulose and polyester. This mat is used for optically testing baby food (artificial milk) for textile fibers.
Bestill nå
SPESIFIKASJONER
Nr. 0048
Vekt 130 g/m²
Environmentally Preferable
Tykkelse 860 µm