Extraction thimbles for DIONEX ASE 100, Whatman™
Leverandør: Whatman products (Cytiva)
10350106 10350108
WHAT10350106EA 2600 NOK
WHAT10350106 WHAT10350108
Extraction thimbles for DIONEX ASE 100, Whatman™
Ekstraktorer Ekstraksjonshylser
Grade 603 standard cellulose extraction thimbles for DIONEX ASE.
  • Ideal for all automated extraction apparatus
  • Maximum temperature: 120 °C
Cellulose material. Grade 603.