pH-indikatorpapir, ruller, Whatman™
Leverandør: Whatman products (Cytiva)
10362030
WHAT10362030EA 370 NOK
WHAT10362030
pH-indikatorpapir, ruller, Whatman™
Testpapir pH-indikatorstrips
Universal indicator papers which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use