Desiccant pack, Whatman™
Leverandør: Whatman products (Cytiva)
10548239
WHAT10548239EA 605 NOK
WHAT10548239
Desiccant pack, Whatman™
Eksikatorer Tørkemidler
Desiccant pack ensure that DNA sampling cards remain dry during transport or storage.
  • High vapour transmission rate
  • Substrate is made with Tyvek® material
  • Non-dusting, strong and hard to tear