Hjelpemidler til askefritt filter, Whatman™
Leverandør: Whatman products (Cytiva)
1703-050
WHAT1703-050EA 2175 NOK
WHAT1703-050
Hjelpemidler til askefritt filter, Whatman™
Filter Filterpapir Kvantitativt filterpapir
Ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive ‘prefilter’ layer on top of normal filter paper.
  • Easily dispersible
  • Powder is of a purity similar to that of ashless quantitative papers
  • Maximum ash content is 0,015%
Leveringsinformasjon: Supplied with two ended scoop for measuring 0,50 or 2,5 g quantities.