Crimp neck vials 11 mm, WHEATON®
Leverandør: DWK Life Sciences
223682
WHEA223682EA 6200 NOK
WHEA223682
Crimp neck vials 11 mm, WHEATON®
Vialer
These vials are made of type I borosilicate glass. They can be used in most autosamplers.
Forpakning: Packaged in shrink-wrapped trays of 100 for convenience and cleanliness.