Screw top vials, WHEATON®
Leverandør: DWK Life Sciences
224795
WHEA224795EA 8175 NOK
WHEA224795
Screw top vials, WHEATON®
Vialer
These vials made of type I borosilicate glass are designed primarily for use in the Waters WISP® 48-position and other autosamplers.
Bestillingsinformasjon: Caps must be ordered separately.
Forpakning: Packaged in shrink- wrapped trays of 100 for convenience and cleanliness.