Dilution vials with cap, WHEATON®
Leverandør: DWK Life Sciences
225002
WHEA225002EA 4525 NOK
WHEA225002
Dilution vials with cap, WHEATON®
Vialer
These vials are manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Marked with a blue line at the 3 ml level
  • Pre-attached solid-top black phenolic screw cap with 14B rubber liner
Forpakning: Vials are packaged in corrugated trays with partitions.