Trypsinising flasks, with screw caps, WHEATON®
Leverandør: DWK Life Sciences
355394 355395 355397 355392 355445 355442 355444
WHEA355394EA 2200 NOK
WHEA355394 WHEA355395 WHEA355397 WHEA355392 WHEA355445 WHEA355442 WHEA355444
Trypsinising flasks, with screw caps, WHEATON®
Kolber Trypsineringskolbe
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Sertifiseringer: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.