Soxhlet Extraction Apparatus
Leverandør: DWK Life Sciences
416005
WHEA416005EA 17850 NOK
WHEA416005
Soxhlet Extraction Apparatus
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
Soxhlet extraction apparatus with standard taper ground joints is used for continuous extraction of analytes from a solid into an organic solvent.
  • Manufactured from low extractable borosilicate glass
As the flask containing the solvent is heated, vapors rise in the larger outside tube, enter the water-cooled condenser and liquate and when the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask.
Sertifiseringer: Conforms to USP Type I and ASTM E 438 Type I, Class A requirements.