Rack for serological pipettes
Leverandør: DWK Life Sciences
72190-0000
WHEA72190-0000EA 16025 NOK
WHEA72190-0000
Rack for serological pipettes
Pipettestativ
Manufactured from heavy-duty clear acrylic. Ideal for the storage of Wheaton serological pipettes from 1 to 100 ml.
Comprises five angled compartments. Top 3 compartments are 3" x 3" and bottom 2 compartments are 3" wide x 1.3/8" high. Also accommodates two pipette controllers on the top - diameter of hole for these is 7/8".