Premium self-refilling repeating syringe with pistol handle grip, Dosys™ 174, WHEATON®
Leverandør: DWK Life Sciences
851372
WHEA851372EA 14350 NOK
WHEA851372
Premium self-refilling repeating syringe with pistol handle grip, Dosys™ 174, WHEATON®
Sprøyter Sprøyter, medisinsk
  • Self-refilling automatic
  • Detachable valve system
  • Enhanced corrosion resistance
Leveringsinformasjon: Supplied with syringe, one meter silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spares for valve and operating instructions.