Coliwasa trumpet, Wheaton®
Leverandør: DWK Life Sciences
885235
WHEA885235EA 8825 NOK
WHEA885235
Coliwasa trumpet, Wheaton®
Prøvetakere Prøvetakere til engangsbruk
This sampler is designed for the extraction of sludge-type or semi-solid materials.
Suitable for boring through materials.