Liquid scintillation vials, without screw cap, WHEATON®
Leverandør: DWK Life Sciences
986532
WHEA986532EA 6800 NOK
WHEA986532
Liquid scintillation vials, without screw cap, WHEATON®
Vialer
Manufactured from low-potassium borosilicate glass to critical manufacturing tolerances, ensuring reliable, consistent results in scintillation counting. Background counts remain low and ultraviolet transmission high. Vials are designed to load properly into scintillation counting equipment without jamming or breaking.
Sertifiseringer: Vials conform to USP Type I and ASTM E 438 Type I, Class A requirements.
Bestillingsinformasjon: Packaged in 5 utility trays with each partitioned tray holding 100 vials. Screw caps can be purchased separately.