Sub surface sampler, Wheaton® Grab Sampler® I
Leverandør: DWK Life Sciences
990250
WHEA990250EA 24725 NOK
WHEA990250
Sub surface sampler, Wheaton® Grab Sampler® I
Prøvetakere Væskeprøvetakere
The sampler features an aluminum body with clamp for securing a sample bottle.
  • Leightweight construction: Golden anodine aluminium tubing and sample bottle with PTFE-lined cap
  • No need to physically enter the sample area
  • Attached extension allows the bottle to be opened and closed under water for discrete sampling
Suitable for sampling in spillways and docks.