WHEATON® Snap On Stopper
Leverandør: DWK Life Sciences
W015258
WHEAW015258EA 0 NOK
WHEAW015258
WHEATON® Snap On Stopper
Propper Gummipropper
Snap-on stopper provides maximum sealing for vials by locking into the blowback ridge.
Provides a tight seal and keeps the stopper from backing out.