Lyophilisation ampoules, Vacule®, WHEATON®
Leverandør: DWK Life Sciences
W651446
WHEAW651446EA 13850 NOK
WHEAW651446
Lyophilisation ampoules, Vacule®, WHEATON®
Ampuller
These lyophilisation ampoules are made of low extractable borosilicate glass.
  • Heat or stopper sealed
Ideal for small volume lyophilization samples.
Sertifiseringer: USP Type , ASTM E 438 Type I, Class A