Classic syringes, Dosys™, WHEATON®
Leverandør: DWK Life Sciences
W852006
WHEAW852006EA 8950 NOK
WHEAW852006
Classic syringes, Dosys™, WHEATON®
Sprøyter Sprøyter, medisinsk
Fast and reliable volume setting, high chemical resistance, and excellent reproducibility make this syringe ideal for laboratories.
  • Automatic refilling
  • Integrated valve system
  • Luer lock nozzle
  • Two-ring and pistol-grip handle
Leveringsinformasjon: Supplied with silicone feed tube, sinker, aspiration and vent cannulas, and operating manual.