NMR sample tubes, thin walled precision, 10 mm Ext.Ø, SP Wilmad-LabGlass
Leverandør: SP Industries
513-7PP-7-5 513-1PS-7-5 513-7PP-8-5 513-5PP-8-5 513-1PP-8-5 513-1PP-7-5 513-3PP-7-5 513-5PP-7-5 513-5PP-9-5
WILM513-7PP-7-5EA 6050 NOK
WILM513-7PP-7-5 WILM513-1PS-7-5 WILM513-7PP-8-5 WILM513-5PP-8-5 WILM513-1PP-8-5 WILM513-1PP-7-5 WILM513-3PP-7-5 WILM513-5PP-7-5 WILM513-5PP-9-5
NMR sample tubes, thin walled precision, 10 mm Ext.Ø, SP Wilmad-LabGlass
Rør NMR-rør
Borosilicate glass, Class A, ASTM Type 1.
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
  • To maximise SNR, Precision NMR tubes have minimal paramagnetic impurities that would impact shimming
  • Tight ID and OD tolerance as small as 0,0065 mm accommodates inserts
  • Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6
NMR PRECISION tubes are manufactured from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438. PRECISION tube has 3 fold less paramagneticMore Product Information