NMR sample tubes, thin walled, PRECISION, 10 mm Ext.Ø, SP Wilmad-LabGlass
Leverandør: SP Industries
513-7PP-7QTZ-5
WILM513-7PP-7QTZ-5EA 18725 NOK
WILM513-7PP-7QTZ-5
NMR sample tubes, thin walled, PRECISION, 10 mm Ext.Ø, SP Wilmad-LabGlass
Rør NMR-rør
Quartz (natural quartz).
  • Possesses a low Boron density as below 0,1 ppm, which guarantees a clean background in Boron-11 NMR studies
  • 2× less dielectric constant than borosilicate glass, which helps improve the quality
  • Not recommended for EPR studies
  • Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6
Extremely low thermal expansion rate and high tensile strength. This feature makes quartz tubes 14 times more robust during cooling/heating process than corresponding Precision glass tubes. Naturally occurring quartzMore Product Information