Transfer pipettes, SP Wilmad-LabGlass
Leverandør: SP Industries
803A
WILM803AEA 3800 NOK
WILM803A
Transfer pipettes, SP Wilmad-LabGlass
Pipetter Pasteurpipetter og overføringspipetter
Glass, class A. These long-tip sample transfer pipettes are ideal for the easy addition or removal of liquid samples from the longest NMR and EPR sample tubes, volumetric flasks with long, narrow necks and chromatography columns.
  • Long tip designed to reach the bottom of 5 mm ext. Ø tube
  • Smooth surface minimises the sample loss
  • Resistant to most organic solvents
  • Transparent body provides easy control on sample loading
  • Free of organic compound contamination
Sertifiseringer: ASTM Type 1