NMR sample tubes, susceptibility matched, Shigemi®, SP Wilmad-LabGlass
Leverandør: SP Industries
BMS-003 BMS-005B
WILMBMS-003EA 8475 NOK
WILMBMS-003 WILMBMS-005B
NMR sample tubes, susceptibility matched, Shigemi®, SP Wilmad-LabGlass
Rør NMR-rør
Susceptibility matched tubes can reduce the sample volume down to 1/3 by minimising the susceptibility gradients occurring at the solvent-air interface.