NMR tube washer, SP Wilmad-LabGlass
Leverandør: SP Industries
WG-1207-J10
WILMWG-1207-J10EA 6150 NOK
WILMWG-1207-J10
NMR tube washer, SP Wilmad-LabGlass
Rør NMR-rør
NMR tube washer is fitted to a filter flask and a inverted sample tube is inserted into the washer, the solvent is introduced into the reservoir using a series of wash bottles. Can perform numerous wash steps and finish by pulling air through the tube.