NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
Leverandør: SP Industries
WG-3000-7-50
WILMWG-3000-7-50EA 3775 NOK
WILMWG-3000-7-50
NMR sample tubes, ECONOMY, 3 mm, SP Wilmad-LabGlass
Rør NMR-rør
High-throughput NMR tubes are made of borosilicate glass Type I, class A.
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments