Septa for Precision NMR tubes, SP Wilmad-LabGlass
Leverandør: SP Industries
WG-3891-100
WILMWG-3891-100EA 2800 NOK
WILMWG-3891-100
Septa for Precision NMR tubes, SP Wilmad-LabGlass
Septum
Natural rubber septa suitable for 5 mm OD thin wall Precision NMR tubes.
  • Designed to maintain excellent glass to rubber contact
  • Suitable for air-sensitive samples, catalytic studies and titration experiments
Colour: white
These septa are manufactured under 'white room' conditions with certified raw materials for consistency.