PILZ® Classic Line LG2 Housing Heating Mantle
Leverandør: WINKLER
W6462-7192
WINKW6462-7192EA 9425 NOK
WINKW6462-7192
PILZ® Classic Line LG2 Housing Heating Mantle
Termostater Varmemantler
Standard metal cased heating mantles.
  • Powder-coated, robust metal housing, with high chemical resistance
  • Hand crocheted heating mantles made of glass silk
Master switch and integrated heating zone switch (heating zone 1 or heating zone 1+2).
Detachable plug-in connection (cable length 0,5 m).
Including personal protection plug with RCD function and cable with Schuko plug type E/F (cable length 1,0 m).